Learn More
BPS Bioscience Inc CD40 - HEK293 Cell Line, 2 vials, Recombinant HEK293 cell line
SDP
Catalog No. NC3930655
Encompass
Recombinant HEK293 cell line expressing full length human CD40 (Tumor necrosis factor receptor superfamily member 5; TNFRSF5), Genbank Accession No. NP_001241.1. Expression is confirmed by real-time qPCR and Western blot.Included with the purchase of these cells is a 10-year license to use the cells and their progeny in your immediate laboratory, for research use only.
Catalog No. NC3930655 Supplier BPS Bioscience Inc Supplier No. 71257
Recombinant HEK293 cell line expressing full length human CD40 (Tumor necrosis factor receptor superfamily member 5; TNFRSF5), Genbank Accession No. NP_001241.1. Expression is confirmed by real-time qPCR and Western blot.Included with the purchase of these cells is a 10-year license to use the cells and their progeny in your immediate laboratory, for research use only.