Learn More
ACROBiosystems ACROBiosystems Human CD40 / TNFRSF5 Protein, His Tag
Catalog No. 501957494
Encompass_Preferred
Recombinant Protein;200UG;Human CD40, His Tag (CD0-H5228) is expressed from human 293 cells (HEK293). It contains AA Glu 21 - Arg 193 (Accession # P25942-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 21.1 kDa. The protein migrates as 29-32 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human CD40, His Tag (Cat. No. CD0-H5228) at 1 μg/mL (100 μL/well) can bind Human CD40 Ligand, Fc Tag (Cat. No. CDL-H5267) with a linear range of 0.2-8 ng/mL (QC tested).
Catalog No. 50-195-7494 Supplier ACROBiosystems Supplier No. CD0H5228200UG
Recombinant Protein;200UG;Human CD40, His Tag (CD0-H5228) is expressed from human 293 cells (HEK293). It contains AA Glu 21 - Arg 193 (Accession # P25942-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 21.1 kDa. The protein migrates as 29-32 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human CD40, His Tag (Cat. No. CD0-H5228) at 1 μg/mL (100 μL/well) can bind Human CD40 Ligand, Fc Tag (Cat. No. CDL-H5267) with a linear range of 0.2-8 ng/mL (QC tested).