Learn More
ACROBiosystems ACROBiosystems Biotinylated Human CD40 / TNFRSF5 Protein, Avitag™,His Tag
Catalog No. 501957403
Encompass_Preferred
Recombinant Protein;25UG;Biotinylated Human CD40, Avitag,His Tag (CD0-H82E8) is expressed from human 293 cells (HEK293). It contains AA Glu 21 - Arg 193 (Accession # P25942-1).;This protein carries an Avi tag (Avitag™) at the C-terminus, followed by a polyhistidine tag. The protein has a calculated MW of 21.8 kDa. The protein migrates as 30-35 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;FACS analysis shows that Biotinylated Human CD40, Avitag,His Tag (Cat. No. CD0-H82E8) can bind to 293T cells overexpressing human CD40L. The concentration of Biotinylated Human CD40 is 3 μg/mL (Routinely tested).
Catalog No. 50-195-7403 Supplier ACROBiosystems Supplier No. CD0H82E825UG
Recombinant Protein;25UG;Biotinylated Human CD40, Avitag,His Tag (CD0-H82E8) is expressed from human 293 cells (HEK293). It contains AA Glu 21 - Arg 193 (Accession # P25942-1).;This protein carries an Avi tag (Avitag™) at the C-terminus, followed by a polyhistidine tag. The protein has a calculated MW of 21.8 kDa. The protein migrates as 30-35 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;FACS analysis shows that Biotinylated Human CD40, Avitag,His Tag (Cat. No. CD0-H82E8) can bind to 293T cells overexpressing human CD40L. The concentration of Biotinylated Human CD40 is 3 μg/mL (Routinely tested).