Learn More
Medchemexpress LLC CD40 Human HEK293 100ug
SDP
Catalog No. 5000180726
Encompass_Preferred
CD40 protein is a TNFSF5/CD40LG receptor that transduces signals through TRAF6 and MAP3K8-mediated pathways activates ERK in macrophages and B cells and induces immunoglobulin secretion CD40 exists as monomers and homodimers and interacts with TRAF proteins (TRAF1 TRAF2 TRAF3 TRAF5 and TRAF6) CD40 Protein Human (HEK293 C-His) is the recombinant human-derived CD40 protein expressed by HEK293 with C-6 His labeled tag
Catalog No. 50-001-80726 Supplier Medchemexpress LLC Supplier No. HYP73499A100UG
CD40 protein is a TNFSF5/CD40LG receptor that transduces signals through TRAF6 and MAP3K8-mediated pathways activates ERK in macrophages and B cells and induces immunoglobulin secretion CD40 exists as monomers and homodimers and interacts with TRAF proteins (TRAF1 TRAF2 TRAF3 TRAF5 and TRAF6) CD40 Protein Human (HEK293 C-His) is the recombinant human-derived CD40 protein expressed by HEK293 with C-6 His labeled tag