Learn More
Medchemexpress LLC CD40 Human HEK293 100ug
SDP
Catalog No. 5000181836
Encompass_Preferred
CD40 protein is a TNFSF5/CD40LG receptor that transduces signals through TRAF6 and MAP3K8-mediated pathways activates ERK in macrophages and B cells and induces immunoglobulin secretion CD40 exists as monomers and homodimers and interacts with TRAF proteins (TRAF1 TRAF2 TRAF3 TRAF5 and TRAF6) CD40 Protein Human (HEK293 Fc) is the recombinant human-derived CD40 protein expressed by HEK293 with C-hFc labeled tag
Catalog No. 50-001-81836 Supplier Medchemexpress LLC Supplier No. HYP72910100UG
CD40 protein is a TNFSF5/CD40LG receptor that transduces signals through TRAF6 and MAP3K8-mediated pathways activates ERK in macrophages and B cells and induces immunoglobulin secretion CD40 exists as monomers and homodimers and interacts with TRAF proteins (TRAF1 TRAF2 TRAF3 TRAF5 and TRAF6) CD40 Protein Human (HEK293 Fc) is the recombinant human-derived CD40 protein expressed by HEK293 with C-hFc labeled tag