Learn More
Enzo Life Sciences CD40 (mouse) monoclonal antibody (FGK45) (preservative free) (100µg)
SDP
Supplier:  Enzo Life Sciences ALX805046PFC100
Encompass_Preferred
Widely used stimulatory MAb to CD40. Shown to indirectly activate natural killer (NK) cells, producing significant antitumor and antimetastatic effects. Effective in boosting immune responses against infectious agents and can potentially be used to treat chronic autoimmune inflammatory processes. Clone: FGK45. Isotype: IgG2a. Immunogen: Recombinant mouse CD40 fusion protein. Source: Purified from hybridoma tissue culture supernatant. Species reactivity: Mouse. Applications: Flow Cytometry, FUNC. Purity: Protein G-affinity purified. Formulation: Liquid. In sterile PBS.
Catalog No. 50-201-0045