Learn More
Enzo Life Sciences CD40 polyclonal antibody (50µg)
SDP
Supplier:  Enzo Life Sciences ADICSS180D
Encompass_Preferred
Host: Rabbit. Immunogen: Synthetic peptide corresponding to 20 residues of human CD40. Source: Purified from rabbit serum. Species reactivity: Human, Mouse, Rat, Sheep, Rabbit, Porcine, Monkey, Hamster, Guinea pig, Dog, Bovine. Applications: WB. Purity: Peptide affinity purified. Formulation: Liquid. In PBS containing 50% glycerol and 0.09% sodium azide.
Catalog No. 50-200-7203