Learn More
Sino Biological Recombinant Human CD40/TNFRSF5 Protein (ECD, hFc & AVI Tag), Biotinylated, HPLC-verified 100 µg
Supplier:  Sino Biological 10774H41HB100
Encompass_Preferred
A DNA sequence encoding the human CD40 ( NP_001241.1) (Met1-Arg193) was expressed with a C-terminal Fc region of human IgG1 tag followed by an AVI tag. The expressed protein was biotinylated in vivo by the Biotin-Protein ligase (BirA enzyme) which is co-expressed.
Catalog No. 50-211-5251