Learn More
Sino Biological Recombinant Human CD40/TNFRSF5 Protein (ECD, mFc Tag)
Supplier:  Sino Biological 10774H38H100
Encompass_Preferred
A DNA sequence encoding the human CD40 (NP_001309350.1) (Met1-Arg193) was expressed with the Fc region of mouse IgG2a at the C-terminus.
Catalog No. 50-211-4865