Learn More
Sino Biological Recombinant Human CD40/TNFRSF5 Protein (ECD, His & AVI Tag), HPLC-verified, Biotinylated
Supplier:  Sino Biological 10774H27HB20
Encompass_Preferred
A DNA sequence encoding the human CD40 (NP_001241.1) (Met1-Arg193) was expressed with a c-terminal polyhistidine tagged AVI tag at the C-terminus. The expressed protein was biotinylated in vivo by the Biotin-Protein ligase (BirA enzyme) which is co-expressed.
Catalog No. 50-211-5233