Learn More
Sino Biological Mouse CD40 / TNFRSF5 Protein (His & AVI Tag), Biotinylated 20ug
Supplier:  Sino Biological 50324M49HB20
Encompass_Preferred
A DNA sequence encoding the mouse CD40 (NP_035741.2) (Met1-Arg193) was expressed with a c-terminal AVI tagged polyhistidine tag at the C-terminus (his-AVI). The expressed protein was biotinylated in vivo by the Biotin-Protein ligase (BirA enzyme) which is co-expressed.
Catalog No. 50-211-5953