Learn More
Enzo Life Sciences CD40 Recombinant monoclonal antibody (G28.5) (Rabbit IgG kappa) (200µg)
SDP
Supplier:  Enzo Life Sciences ENZABS4470200
Encompass_Preferred
Clone: G28.5. Isotype: IgGκ. Immunogen: Human erythrocyte-rosette-negative tonsillar lymphocytes. Species reactivity: Human. Applications: Flow Cytometry, IHC, IP. Purity: Protein A affinity purified. Formulation: Liquid. In PBS containing 0.02% Proclin 300.
Catalog No. 50-201-2974