Learn More
Enzo Life Sciences CD40, soluble (human) ELISA kit (1x96wells)
SDP
Supplier:  Enzo Life Sciences ALX850262KI01
Encompass_Preferred
The CD40, soluble (human) ELISA kit is a complete assay for the quantitative determination of human soluble CD40 concentration. This kit is to be used with cell culture supernatants, serum and other body fluids from human origin. Alternative name: TNFRSF 5. Assay Time: <3.25 hours. Applications: ELISA . Species reactivity: Human .
Catalog No. 50-201-0144