Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ CD40/TNFRSF5 Antibody Pair [HRP]
SDP

Catalog No. NB009627
Change view
Click to view available options
Quantity:
15 Plates
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB009627 15 Plates
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB009627 Supplier R&D Systems™ Supplier No. NBP27929415PLATES

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

A Novus product, part of R&D Systems. Monoclonal Antibody Pair

specifically detects in Human samples. It is validated for ELISA, Sandwich ELISA.

Specifications

Assay Range 3.9 pg/mL
Assay Sensitivity 3.91-250 pg/mL
Assay Sandwich ELISA
Conjugate HRP
Product Line Novus
Product Type Antibody Pairs
Specificity This solid phase sandwich ELISA utilizes a monoclonal antibody specific for Human CD40/TNFRSF5.
Target Species Human
For Use With (Application) Sandwich ELISA
Gene Alias B cell surface antigen CD40, B-cell surface antigen CD40, Bp50B cell-associated molecule, CD40 antigen, CD40 molecule, TNF receptor superfamily member 5, CD40 type II isoform, CD40L receptor, CDw40, MGC9013, nerve growth factor receptor-related B-lymphocyte activation molecule, p50, TNFRSF5CD40 antigen (TNF receptor superfamily member 5), tumor necrosis factor receptor superfamily member 5, tumor necrosis factor receptor superfamily, member 5
Gene ID (Entrez) 958
Gene Symbol CD40
Host Species Rabbit/Mouse
Kit Contents
  • Standard: Each vial contains 20 ng of recombinant CD40/TNFRSF5.
  • Capture Antibody: 1.0 mg/mL of mouse anti-CD40/TNFRSF5 monoclonal antibody.
  • Detection Antibody: 0.5 mg/mL rabbit anti-CD40/TNFRSF5 monoclonal antibody conjugated to horseradish-peroxidase (HRP).
  • Quantity 15 Plates
    Regulatory Status RUO
    Research Discipline Adaptive Immunity, Apoptosis, Asthma, B Cell Development and Differentiation Markers, Cancer, Hematopoietic Stem Cell Markers, Immunology, Signal Transduction, Stem Cell Markers
    Storage Requirements Storage is content dependent.
    Target CD40/TNFRSF5
    Technique 1. Capture Antibody: Dilute to a working concentration of 2.0 μg/mL in CBS before coating 2. Detection Antibody: Dilute to working concentration of 0.5 μg/mL in detection antibody dilution buffer before use. 3. Standard: Reconstitute with 1 ml detection antibody dilution buffer. After reconstitution, store at -20 to -80°C in a manual defrost freezer. A seven-point standard curve usi ng 2-fold serial dilutions in sample dilution buffer, and a high standard of 250 pg/mL is recommended
    Show More Show Less
    Product Title
    Select an issue

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.