Learn More
Genscript Corporation CD40/TNFRSF5 His Mouse
Catalog No. 5000058489
Encompass
CD40 is a costimulatory protein found on antigen presenting cells and is required for their activation The binding of CD154 (CD40L) on TH cells to CD40 activates antigen presenting cells and induces a variety of downstream effects CD40 molecule is a potential target for cancer immunotherapy There are number of completed and ongoing clinical trials where agonistic anti-CD40 monoclonal antibodies are employed to activate an anti-tumor T cell response via activation of dendritic cells
Catalog No. 50-000-58489 Supplier Genscript Corporation Supplier No. Z04319100
CD40 is a costimulatory protein found on antigen presenting cells and is required for their activation The binding of CD154 (CD40L) on TH cells to CD40 activates antigen presenting cells and induces a variety of downstream effects CD40 molecule is a potential target for cancer immunotherapy There are number of completed and ongoing clinical trials where agonistic anti-CD40 monoclonal antibodies are employed to activate an anti-tumor T cell response via activation of dendritic cells