Learn More
ACROBiosystems Recombinant Protein;Biotinylated Human CD40 / TNFRSF5, Fc Tag & Avi Tag (Avitag™);HEK293;200UG;TN5,CD40,CD-40,Bp50,Bp-50,CDW40,MGC9013,TNFRSF5,TNFRSF-5,p50,p-50
Catalog No. 501658315
Encompass_Preferred
Recombinant Protein;Biotinylated Human CD40 / TNFRSF5, Fc Tag & Avi Tag (Avitag™);200UG;Biotinylated Human CD40, Fc Tag (TN5-H82F9) is expressed from human 293 cells (HEK293). It contains AA Glu 21 - Arg 193 (Accession # P25942-1).;This protein carries a human IgG1 Fc tag at the C-terminus, followed by a Avi tag (Avitag™). The protein has a calculated MW of 48.1 kDa. The protein migrates as 60-66 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human CD40 Ligand, His Tag (Cat. No. CDL-H5248) at 5 μg/mL (100 μL/well) can bind Biotinylated Human CD40, Fc Tag (Cat. No. TN5-H82F9) with a linear range of 0.313-2.5 μg/mL (QC tested).
Catalog No. 50-165-8315 Supplier ACROBiosystems Supplier No. TN5H82F9200UG
Recombinant Protein;Biotinylated Human CD40 / TNFRSF5, Fc Tag & Avi Tag (Avitag™);200UG;Biotinylated Human CD40, Fc Tag (TN5-H82F9) is expressed from human 293 cells (HEK293). It contains AA Glu 21 - Arg 193 (Accession # P25942-1).;This protein carries a human IgG1 Fc tag at the C-terminus, followed by a Avi tag (Avitag™). The protein has a calculated MW of 48.1 kDa. The protein migrates as 60-66 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human CD40 Ligand, His Tag (Cat. No. CDL-H5248) at 5 μg/mL (100 μL/well) can bind Biotinylated Human CD40, Fc Tag (Cat. No. TN5-H82F9) with a linear range of 0.313-2.5 μg/mL (QC tested).