Learn More
ACROBiosystems Recombinant Protein;Mouse CD40 / TNFRSF5 Protein;HEK293;100UG;TN5,CD40,CD-40,Bp50,Bp-50,CDW40,MGC9013,TNFRSF5,TNFRSF-5,p50,p-50
Catalog No. 501658743
Encompass_Preferred
Recombinant Protein;Mouse CD40 / TNFRSF5 Protein;100UG;Mouse CD40, His Tag (TN5-M52H8) is expressed from human 293 cells (HEK293). It contains AA Val 24 - Arg 193 (Accession # P27512).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 20.8 kDa. The protein migrates as 30 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Mouse CD40, His Tag (Cat. No. TN5-M52H8) at 5μg/mL (100 μL/well) can bind Mouse CD40 Ligand, Fc Tag (Cat. No. CDL-M526x) with a linear range of 1-16 ng/mL (QC tested).
Catalog No. 50-165-8743 Supplier ACROBiosystems Supplier No. TN5M52H8100UG
Recombinant Protein;Mouse CD40 / TNFRSF5 Protein;100UG;Mouse CD40, His Tag (TN5-M52H8) is expressed from human 293 cells (HEK293). It contains AA Val 24 - Arg 193 (Accession # P27512).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 20.8 kDa. The protein migrates as 30 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Mouse CD40, His Tag (Cat. No. TN5-M52H8) at 5μg/mL (100 μL/well) can bind Mouse CD40 Ligand, Fc Tag (Cat. No. CDL-M526x) with a linear range of 1-16 ng/mL (QC tested).