Learn More
ACROBiosystems Recombinant Protein;Rhesus macaque CD40 / TNFRSF5 Protein;HEK293;1MG;CD0,CD40,CD-40,Bp50,Bp-50,CDW40,MGC9013,TNFRSF5,TNFRSF-5,p50,p-50
Catalog No. 501658594
Encompass_Preferred
Recombinant Protein;Rhesus macaque CD40 / TNFRSF5 Protein;1MG;Rhesus macaque CD40, His Tag (CD0-C52H7) is expressed from human 293 cells (HEK293). It contains AA Glu 21 - Arg 193 (Accession # NP_001252791.1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 21.1 kDa. The protein migrates as 30-34 kDa under reducing (R) condition (SDS-PAGE).
Catalog No. 50-165-8594 Supplier ACROBiosystems Supplier No. CD0C52H71MG
Recombinant Protein;Rhesus macaque CD40 / TNFRSF5 Protein;1MG;Rhesus macaque CD40, His Tag (CD0-C52H7) is expressed from human 293 cells (HEK293). It contains AA Glu 21 - Arg 193 (Accession # NP_001252791.1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 21.1 kDa. The protein migrates as 30-34 kDa under reducing (R) condition (SDS-PAGE).