Learn More
ACROBiosystems Recombinant Protein;Cynomolgus CD40 / TNFRSF5 Protein;HEK293;1MG;CD0,CD40,CD-40,Bp50,Bp-50,CDW40,MGC9013,TNFRSF5,TNFRSF-5,p50,p-50
Catalog No. 501659121
Encompass_Preferred
Recombinant Protein;Cynomolgus CD40 / TNFRSF5 Protein;1MG;Cynomolgus CD40, His Tag (CD0-C52H6) is expressed from human 293 cells (HEK293). It contains AA Glu 21 - Arg 193 (Accession # G7PG38).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 21.1 kDa. The protein migrates as 30-34 kDa under reducing (R) condition (SDS-PAGE).;Immobilized Cynomolgus CD40, His Tag (Cat. No. CD0-C52H6) at 5μg/mL (100 μL/well) can bind Mouse CD40 Ligand, Fc Tag (Cat. No. CDL-M526x) with a linear range of 1-6.6 ng/mL (QC tested).
Catalog No. 50-165-9121 Supplier ACROBiosystems Supplier No. CD0C52H61MG
Recombinant Protein;Cynomolgus CD40 / TNFRSF5 Protein;1MG;Cynomolgus CD40, His Tag (CD0-C52H6) is expressed from human 293 cells (HEK293). It contains AA Glu 21 - Arg 193 (Accession # G7PG38).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 21.1 kDa. The protein migrates as 30-34 kDa under reducing (R) condition (SDS-PAGE).;Immobilized Cynomolgus CD40, His Tag (Cat. No. CD0-C52H6) at 5μg/mL (100 μL/well) can bind Mouse CD40 Ligand, Fc Tag (Cat. No. CDL-M526x) with a linear range of 1-6.6 ng/mL (QC tested).