Learn More
Medchemexpress LLC CD40L/CD154/TRAP Hu 100ug
SDP
Catalog No. 5000196003
Encompass_Preferred
Recombinant human CD40L soluble form (M113-L261) produced in HEK293 cells with N-terminal hFc and Flag tags, supplied as a lyophilized powder. The product is high purity and low endotoxin, formulated with trehalose to support stability and performance in functional binding and cell-based assays.
  • High purity (>95%) for reliable biochemical assays.
  • Low endotoxin level (<1 EU/μg) to minimize cellular activation artifacts.
  • N-terminal hFc and Flag tags facilitate detection and capture.
  • Lyophilized formulation with trehalose for enhanced stability.
  • Validated in ELISA, LSPR, and B-cell proliferation assays.
  • Stable when stored at -20°C and suitable for long-term storage with carrier protein.
Catalog No. 50-001-96003 Supplier Medchemexpress LLC Supplier No. HYP72028100UG
Recombinant human CD40L soluble form (M113-L261) produced in HEK293 cells with N-terminal hFc and Flag tags, supplied as a lyophilized powder. The product is high purity and low endotoxin, formulated with trehalose to support stability and performance in functional binding and cell-based assays.
  • High purity (>95%) for reliable biochemical assays.
  • Low endotoxin level (<1 EU/μg) to minimize cellular activation artifacts.
  • N-terminal hFc and Flag tags facilitate detection and capture.
  • Lyophilized formulation with trehalose for enhanced stability.
  • Validated in ELISA, LSPR, and B-cell proliferation assays.
  • Stable when stored at -20°C and suitable for long-term storage with carrier protein.