Learn More
AdipoGen CD40L (human) (multimeric) (rec.) (Biotin) (3 x 10 µg)
SDP
Supplier:  AdipoGen AG40B0010B3010
Encompass_Preferred
Recombinant protein. Human CD40L (aa 116-261) is fused at the N-terminus to mouse ACRP30headless (aa 18-111) and a FLAG-tag. Source/Host: CHO cells. Purity: >95% (SDS-PAGE). Lyophilized. Contains PBS. MultimericCD40L™ is a high activity construct in which two trimeric CD40 ligands are artificially linked via the collagen domain of ACRP30. This construct very effectively simulates the natural membrane-assisted aggregation of CD40L in vivo. It provides a simple and equally potent alternative to [CD40L+enhancer] combinations. MultimericCD40L™ has shown to suppress alum-induced IL-1beta release and caspase-1 activation in a dose-, CD40- and time dependent manner, without affecting BMDM viability. It also effectively suppressed the inflammasome function triggered by NLRP3 activators. The secretion of caspase-1 independent inflammatory mediators has been shown to be unaltered or even enhanced.
Catalog No. 50-205-3079