Learn More
Enzo Life Sciences CD40L (human) monoclonal antibody (blocking) (MK13A4) (100 µg)
SDP
Supplier:  Enzo Life Sciences ALX805037C100
Encompass_Preferred
Clone: MK13A4. Host: Mouse. Isotype: IgG1. Immunogen: Recombinant human CD40L (CD154). UniProt: P29965. Species Reactivity: Human. Applications: Flow Cytometry. FUNC. Formulation: Liquid. In PBS, pH 7.2-7.4. Handling: Avoid freeze/thaw cycles. After opening, prepare aliquots and store at -20°C.
Catalog No. NC1188218