Learn More
Enzo Life Sciences CD40L (mouse) monoclonal antibody (MR1) (100µg)
SDP
Supplier:  Enzo Life Sciences ALX804331C100
Encompass_Preferred
Clone: MR1. Isotype: IgG1. Immunogen: Mouse T-cells (Th1). Species reactivity: Mouse. Applications: FUNC. Purity: Protein A-affinity purified. Formulation: Liquid. In PBS.
Catalog No. 50-200-9868