Learn More
AdipoGen CD40L (mouse) (multimeric) (rec.) (3 x 10 µg)
SDP
Supplier:  AdipoGen AG40B00203010
Encompass_Preferred
Protein. Mouse CD40L (aa 115-260) is fused at the N-terminus to mouse ACRP30headless (aa 18-111) and a FLAG-tag. Source: CHO cells. Endotoxin content: <0.01EU/µg purified protein (LAL test; Lonza). Lyophilized. Contains PBS. Binds to human and mouse CD40. Purity: >95% (SDS-PAGE). MultimericCD40L™ is a high activity construct in which two trimeric CD40 ligands are artificially linked via the collagen domain of ACRP30. This construct very effectively simulates the natural membrane-assisted aggregation of CD40L in vivo. It provides a simple and equally potent alternative to CD40L+enhancer combinations. MultimericCD40L™ has shown to suppress alum-induced IL-1beta release and caspase-1 activation in a dose-, CD40- and time dependent manner, without affecting BMDM (Bone marrow-derived macrophages) viability. It also effectively suppressed the inflammasome function triggered by NLRP3 activators.
Catalog No. 50-205-3091