Learn More
AdipoGen CD40L ms multimeric Biot
SDP
Supplier:  AdipoGen AG40B0020B3010
Encompass_Preferred
Recombinant protein. Mouse CD40L aa 115-260 is fused at the N-terminus to mouse ACRP30headless aa 18-111 and a FLAG -tag. Binds to human and mouse CD40. Endotoxin <0.05EU/µg purified protein LAL test Lonza. Lyophilized. Contains PBS. The costimulatory molecule CD40, a member of the tumor necrosis factor TNF receptor superfamily, critically regulates B cell and T cell function in adaptive immunity and inflammation by interacting with CD40L CD154. CD40L mediates a range of activities on B cells, including induction of activation-associated surface antigen, entry into cell cycle, isotype switching, immunoglobulin secretion and memory generation. CD40-CD40L interaction also plays important roles in monocyte activation and DC maturation. MultimericCD40L™ is a high activity construct in which two trimeric CD40 ligands are artificially linked via the collagen domain of ACRP30. This construct very effectively simulates the natural membrane-assisted aggregation of CD40L in vivo.
Catalog No. 50-256-2934