Learn More
Enzo Life Sciences CD40L Recombinant monoclonal antibody (hu5c8) (Rabbit IgG kappa) (200µg)
SDP
Supplier:  Enzo Life Sciences ENZABS4490200
Encompass_Preferred
Clone: hu5c8 (Ruplizumab). Isotype: IgGκ. Immunogen: Human CD40L. Species reactivity: Human. Specificity: The antibody binds to CD40L with an IC50 of 0.845 µg/ml. Applications: Flow Cytometry, WB, FUNC. Purity: Protein A affinity purified. Formulation: Liquid. In PBS containing 0.02% Proclin 300.
Catalog No. 50-201-2976