Learn More
Enzo Life Sciences CD40L (soluble) (mouse), (recombinant) (10µg)
SDP
Catalog No. 502009403
Encompass_Preferred
Alternative name: CD154, TNFSF 5, gp39. Concentration: 0.1mg/ml after reconstitution. Purity: ≥90% (SDS-PAGE). Formulation: Lyophilized. Contains PBS. Source: Produced in HEK 293 cells. The extracellular domain of mouse CD40L (CD154) (aa 115-260) is fused at the N-terminus to a linker peptide (8 aa) and a FLAG-tag. Specificity: Binds to human and mouse CD40. Long Term Storage: -20°C. Applications: ELISA .
Catalog No. 50-200-9403 Supplier Enzo Life Sciences Supplier No. ALX522070C010
Alternative name: CD154, TNFSF 5, gp39. Concentration: 0.1mg/ml after reconstitution. Purity: ≥90% (SDS-PAGE). Formulation: Lyophilized. Contains PBS. Source: Produced in HEK 293 cells. The extracellular domain of mouse CD40L (CD154) (aa 115-260) is fused at the N-terminus to a linker peptide (8 aa) and a FLAG-tag. Specificity: Binds to human and mouse CD40. Long Term Storage: -20°C. Applications: ELISA .