Learn More
Enzo Life Sciences CD40L (soluble) (human), (recombinant) (10 µg)
SDP
Supplier:  Enzo Life Sciences ALX5220156010
Encompass_Preferred
Increases the stimulation of B cells. Alternative name: CD154, TNFSF 5, gp39. MW: ~18kDa (SDS-PAGE). Concentration: 0.1mg/ml after reconstitution. Endotoxin content: E. coli. The extracellular domain of human CD40L (CD154) (aa 116-261) is fused at the N-terminus to a linker peptide (6 aa) and a FLAG-tag. Specificity: Binds to human CD40. Use/Stability: Stable for at least 6 months after receipt when stored at -20°C. UniProt: P29965. Application: ELISA. Handling: Avoid freeze/thaw cycles. After reconstitution, prepare aliquots and store at -20°C. Long Term Storage: -20°C
Catalog No. NC9450819