Learn More
Aviva Systems Biology Corp CD40LG Antibody - middle region (ARP33831_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP33831_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CD40LG. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human CD40LG. Alternate names: CD154, CD40L, HIGM1, IGM, IMD3, T-BAM, TNFSF5, TRAP, gp39, hCD40LApplicationIHC, WB. Species reactivity: Cow, Dog, Horse, Human, Mouse, Pig, Rabbit, Rat, Sheep
Catalog No. 50-014-55164