Learn More
ACROBiosystems ACROBiosystems Mouse CD40 Ligand / TNFSF5 Protein, Fc Tag
Supplier:  ACROBiosystems CDLM526X1MG
Encompass_Preferred
Recombinant Protein;1MG;Mouse CD40 Ligand, Fc Tag (CDL-M526x) is expressed from human 293 cells (HEK293). It contains AA Gly 115 - Leu 260 (Accession # P27548-1).;This protein carries a human IgG1 Fc tag at the N-terminus. The protein has a calculated MW of 42.9 kDa. As a result of glycosylation, the protein migrates as 52 kDa under reducing (R) condition, and 90-116 kDa under non-reducing (NR) condition (SDS-PAGE).;Immobilized Mouse CD40, His Tag (Cat. No. TN5-M52H8) at 5 μg/mL (100 μL/well) can bind Mouse CD40 Ligand, Fc Tag (Cat. No. CDL-M526x) with a linear range of 1-16 ng/mL (QC tested).
Catalog No. 50-195-7344