Learn More
ACROBiosystems ACROBiosystems Mouse CD40 Ligand / TNFSF5 Protein, His Tag (active trimer) (MALS verified)
Catalog No. 501957289
Encompass_Preferred
Recombinant Protein;1MG;Mouse CD40 Ligand, His Tag (active trimer) (MALS verified) (CDL-M5248) is expressed from human 293 cells (HEK293). It contains AA Gly 115 - Leu 260 (Accession # P27548-1).;This protein carries a polyhistidine tag at the N-terminus. The protein has a calculated MW of 16.8 kDa. The protein migrates as 21-25 kDa under reducing (R) condition (SDS-PAGE).;Immobilized Mouse CD40, Fc Tag (Cat. No. TN5-M5259) at 2 μg/mL ( 100 μl/well ) can bind Mouse CD40 Ligand, His Tag (active trimer) (MALS verified) (Cat. No. CDL-M5248) with a linear range of 5-20 ng/mL (QC tested).
Catalog No. 50-195-7289 Supplier ACROBiosystems Supplier No. CDLM52481MG
Recombinant Protein;1MG;Mouse CD40 Ligand, His Tag (active trimer) (MALS verified) (CDL-M5248) is expressed from human 293 cells (HEK293). It contains AA Gly 115 - Leu 260 (Accession # P27548-1).;This protein carries a polyhistidine tag at the N-terminus. The protein has a calculated MW of 16.8 kDa. The protein migrates as 21-25 kDa under reducing (R) condition (SDS-PAGE).;Immobilized Mouse CD40, Fc Tag (Cat. No. TN5-M5259) at 2 μg/mL ( 100 μl/well ) can bind Mouse CD40 Ligand, His Tag (active trimer) (MALS verified) (Cat. No. CDL-M5248) with a linear range of 5-20 ng/mL (QC tested).