Learn More
ACROBiosystems ACROBiosystems Human CD40 Ligand / TNFSF5 Protein, His,Flag Tag (active trimer) (MALS verified)
Catalog No. 501956437
Encompass_Preferred
Recombinant Protein;1MG;Human CD40 Ligand, His,Flag Tag (active trimer) (MALS verified) (CDL-H52Db) is expressed from human 293 cells (HEK293). It contains AA Gly 116 - Leu 261 (Accession # P29965-1) trimer Design.;This protein carries a polyhistidine tag at the N-terminus, followed by a flag tag. The protein has a calculated MW of 49.9 kDa. The protein migrates as 45-55 kDa under non-reducing (NR) condition (SDS-PAGE).;Immobilized Human CD40, Fc Tag (Cat. No. CD0-H5253) at 5 μg/mL (100 μL/well) can bind Human CD40 Ligand, His,Flag Tag (active trimer) (MALS verified) (Cat. No. CDL-H52Db) with a linear range of 20-156 ng/mL (QC tested).
Catalog No. 50-195-6437 Supplier ACROBiosystems Supplier No. CDLH52DB1MG
Recombinant Protein;1MG;Human CD40 Ligand, His,Flag Tag (active trimer) (MALS verified) (CDL-H52Db) is expressed from human 293 cells (HEK293). It contains AA Gly 116 - Leu 261 (Accession # P29965-1) trimer Design.;This protein carries a polyhistidine tag at the N-terminus, followed by a flag tag. The protein has a calculated MW of 49.9 kDa. The protein migrates as 45-55 kDa under non-reducing (NR) condition (SDS-PAGE).;Immobilized Human CD40, Fc Tag (Cat. No. CD0-H5253) at 5 μg/mL (100 μL/well) can bind Human CD40 Ligand, His,Flag Tag (active trimer) (MALS verified) (Cat. No. CDL-H52Db) with a linear range of 20-156 ng/mL (QC tested).