Learn More
ACROBiosystems Recombinant Protein;Biotinylated Human CD40 Ligand / TNFSF5 Protein, Fc Tag, Avi Tag (Avitag™);HEK293;200UG;CDL,CD40LG,CD154,CD40L,HIGM1,IGM,IMD3,T-BAM,TNFSF5,TRAP,gp39,CD40 Ligand,CD40-LG,CD40 L
Supplier:  ACROBiosystems CDLH82F1200UG
Encompass_Preferred
Recombinant Protein;Biotinylated Human CD40 Ligand / TNFSF5 Protein, Fc Tag, Avi Tag (Avitag™);200UG;Biotinylated Human CD40 Ligand, Fc Tag (CDL-H82F1) is expressed from human 293 cells (HEK293). It contains AA Met 113 - Leu 261 (Accession # P29965-1).;This protein carries an Avi tag (Avitag™) at the N-terminus, followed by a human IgG1 Fc tag. The protein has a calculated MW of 45.0 kDa. The protein migrates as 50-55 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human CD40, His Tag (Cat. No. CD0-H5228) at 2 μg/mL (100 μL/well) can bind Biotinylated Human CD40 Ligand, Fc Tag (Cat. No. CDL-H82F1) with a linear range of 1-31 ng/mL (QC tested).
Catalog No. 50-165-8219