Learn More
ACROBiosystems Recombinant Protein;Human / Rhesus macaque CD40 Ligand / TNFSF5 Protein, Mouse IgG2a Fc Tag, low endotoxin;HEK293;100UG;CDL,CD40LG,CD154,CD40L,HIGM1,IGM,IMD3,T-BAM,TNFSF5,TRAP,gp39,CD40 Ligand,CD40-LG,CD40 L
Catalog No. 501658030
Encompass_Preferred
Recombinant Protein;Human / Rhesus macaque CD40 Ligand / TNFSF5 Protein, Mouse IgG2a Fc Tag, low endotoxin;100UG;Human / Rhesus macaque CD40 Ligand, Mouse IgG2a Fc Tag, low endotoxin (CDL-H5256) is expressed from human 293 cells (HEK293). It contains AA Met 113 - Leu 261 (Accession # P29965-1).;This protein carries a mouse IgG2a Fc tag at the N-terminus. The protein has a calculated MW of 43.1 kDa. The protein migrates as 45-50 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human CD40, His Tag (Cat. No. CD0-H5228) at 1 μg/mL (100 μL/well) can bind Human / Rhesus macaque CD40 Ligand, Mouse IgG2a Fc Tag (Cat. No. CDL-H5256) with a linear range of 0.4-6 ng/mL (QC tested).
Catalog No. 50-165-8030 Supplier ACROBiosystems Supplier No. CDLH5256100UG
Recombinant Protein;Human / Rhesus macaque CD40 Ligand / TNFSF5 Protein, Mouse IgG2a Fc Tag, low endotoxin;100UG;Human / Rhesus macaque CD40 Ligand, Mouse IgG2a Fc Tag, low endotoxin (CDL-H5256) is expressed from human 293 cells (HEK293). It contains AA Met 113 - Leu 261 (Accession # P29965-1).;This protein carries a mouse IgG2a Fc tag at the N-terminus. The protein has a calculated MW of 43.1 kDa. The protein migrates as 45-50 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human CD40, His Tag (Cat. No. CD0-H5228) at 1 μg/mL (100 μL/well) can bind Human / Rhesus macaque CD40 Ligand, Mouse IgG2a Fc Tag (Cat. No. CDL-H5256) with a linear range of 0.4-6 ng/mL (QC tested).