Learn More
ACROBiosystems Recombinant Protein;Human CD40 / TNFRSF5 Protein, Mouse IgG2a Fc Tag, low endotoxin;HEK293;1MG;CD0,CD40,CD-40,Bp50,Bp-50,CDW40,MGC9013,TNFRSF5,TNFRSF-5,p50,p-50
Catalog No. 501657994
Encompass_Preferred
Recombinant Protein;Human CD40 / TNFRSF5 Protein, Mouse IgG2a Fc Tag, low endotoxin;1MG;Human CD40, Mouse IgG2a Fc Tag, low endotoxin (CD0-H525a) is expressed from human 293 cells (HEK293). It contains AA Glu 21 - Arg 193 (Accession # P25942-1).;This protein carries a mouse IgG2a Fc tag at the C-terminus. The protein has a calculated MW of 46.1 kDa. The protein migrates as 58 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human CD40 Ligand, Fc Tag (Cat. No. CDL-H5267) at 5 μg/mL (100 μL/well) can bind Human CD40, Mouse IgG2a Fc Tag, low endotoxin (Cat. No. CD0-H525a) with a linear range of 20-156 ng/mL (QC tested).
Catalog No. 50-165-7994 Supplier ACROBiosystems Supplier No. CD0H525A1MG
Recombinant Protein;Human CD40 / TNFRSF5 Protein, Mouse IgG2a Fc Tag, low endotoxin;1MG;Human CD40, Mouse IgG2a Fc Tag, low endotoxin (CD0-H525a) is expressed from human 293 cells (HEK293). It contains AA Glu 21 - Arg 193 (Accession # P25942-1).;This protein carries a mouse IgG2a Fc tag at the C-terminus. The protein has a calculated MW of 46.1 kDa. The protein migrates as 58 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human CD40 Ligand, Fc Tag (Cat. No. CDL-H5267) at 5 μg/mL (100 μL/well) can bind Human CD40, Mouse IgG2a Fc Tag, low endotoxin (Cat. No. CD0-H525a) with a linear range of 20-156 ng/mL (QC tested).