Learn More
Enzo Life Sciences CD42c (human) monoclonal antibody (Gi27) (100µg)
SDP
Supplier:  Enzo Life Sciences ALX805026C100
Encompass_Preferred
Clone: Gi27. Isotype: IgG2b. Immunogen: Human platelets. Species reactivity: Human. Specificity: Recognizes the GPIbβ subunit (CD42c) of the GPIb/IX/V (CD42a/CD42b/CD42c/CD42d) complex. Applications: IP, WB. Formulation: Liquid. 100µg of purified antibody in 200µl PBS containing 0.05% sodium azide.
Catalog No. 50-201-0037