Learn More
Biocare Medical, LLC CD43 Antibody - Concentrate, 1.0 ml
SDP
Supplier:  Biocare Medical, LLC CM005C
Encompass_Preferred
CD43 antibody recognizes a 95/115/135kDa (depending upon the extent of glycosylation) cell surface glycoprotein, identified as CD43 (leukosialin, sialophorin, or leukocyte sialoglycoprotein) CD43 is shown to be expressed on all thymocytes, T-cells and in endothelial cells. CD43 may also aid in distinguishing extranodal marginal zone B-cell lymphoma from other reactive processes in the skin. The CD43 antibody has also been shown to be useful in aiding in the identification and classification of T-cell malignancies and low-grade B-cell lymphomas.
Catalog No. 50-824-12