Learn More
enQuire Bioreagents Calponin-1 Anti-Human Monoclonal [Clone SPM169], Size: 100ug
Supplier:  enQuire Bioreagents 6693MSM2P1
Encompass_Preferred
Product Type: Antibody. Application: Flow, IF, WB, IHC. Conjugate/Tag: Unconjugated. Clone: SPN/839. Clonality: Monoclonal. Host: Mouse. Isotype: IgG1 kappa. Positive Control: Paracortex in a tonsil or a reactive lymph node. Cellular Staining: Cell surface. Buffer: 10mM PBS with 0.05% BSA & 0.05% azide. Concentration: 200ug/ml. Purity: Protein A/G Purified. Immunogen: Recombinant full-length human SPN protein. Storage: Short term: Store at 2-4C. Long term, aliquot and store at -20C. MW: 95, 115, or 135kDa.Chromosome: 16p11.2. Gene Symbol: SPN. Gene ID: 6693. Ensemble: ENSG00000197471. Uniprot: NP_003114.
Catalog No. 50-188-5186