Learn More
Genscript Corporation CD43 hFc Chimera Mouse
Catalog No. 5000056628
Encompass
CD43 is a large transmembrane protein involved in T cell activation Previous studies of CD43-/- mice in viral models have demonstrated a role for CD43 in Th1/Th2 skewing activation of Foxp3 Treg and T cell apoptosis CD43 (leukosialin) is a large sialoglycoprotein abundantly expressed on the surface of most cells from the hematopoietic lineage CD43 is directly involved in the contact between cells participating in a series of events such as signaling adherence and host parasite interactions
Catalog No. 50-000-56628 Supplier Genscript Corporation Supplier No. Z04320100
CD43 is a large transmembrane protein involved in T cell activation Previous studies of CD43-/- mice in viral models have demonstrated a role for CD43 in Th1/Th2 skewing activation of Foxp3 Treg and T cell apoptosis CD43 (leukosialin) is a large sialoglycoprotein abundantly expressed on the surface of most cells from the hematopoietic lineage CD43 is directly involved in the contact between cells participating in a series of events such as signaling adherence and host parasite interactions