Learn More
Genscript Corporation CD43 His Cynomolgus
Supplier:  Genscript Corporation Z04121500
Encompass
CD43 (leukosialin) is a large sialoglycoprotein abundantly expressed on the surface of most cells from the hematopoietic lineage CD43 is directly involved in the contact between cells participating in a series of events such as signaling adherence and host parasite interactions The induction of acute myocarditis involves the engagement of CD43 cytoplasmic tripeptide sequence KRR to ezrin-radixin-moiesin cytoskeletal proteins
Catalog No. 50-000-58968