Learn More
Medchemexpress LLC CD43 Human HEK293 20ug | 20ug
SDP
Catalog No. 5000187656
Encompass_Preferred
CD43 protein is an important leukocyte surface sialic acid protein that plays a key role in T cell regulation It has a positive impact on T cell function including activation proliferation differentiation trafficking and migration especially by helping T cells migrate to lymph nodes through ERM proteins CD43 Protein Human (HEK293 His) is the recombinant human-derived CD43 protein expressed by HEK293 with C-8 His labeled tag
Catalog No. 50-001-87656 Supplier Medchemexpress LLC Supplier No. HYP7788720UG
CD43 protein is an important leukocyte surface sialic acid protein that plays a key role in T cell regulation It has a positive impact on T cell function including activation proliferation differentiation trafficking and migration especially by helping T cells migrate to lymph nodes through ERM proteins CD43 Protein Human (HEK293 His) is the recombinant human-derived CD43 protein expressed by HEK293 with C-8 His labeled tag