Learn More
Medchemexpress LLC CD43 protein (mouse), Fc fusion, expressed in HEK293 cells | 95.0% | 5 UG
SDP
Catalog No. 5000291277
Encompass_Preferred
Recombinant mouse CD43 (leukosialin) extracellular domain produced in HEK293 cells and supplied as an Fc fusion protein. CD43 plays a regulatory role in T cell activation, proliferation, differentiation, trafficking, and migration. The product is supplied as a 5 μg aliquot with reported purity of 95% and an apparent molecular weight of ~72-120 kDa.
  • Recombinant extracellular domain of mouse CD43.
  • Expressed in HEK293 cells for mammalian post-translational modifications.
  • Fc fusion tag for improved stability and detection.
  • Reported purity of 95.0%.
  • Apparent molecular weight approximately 72-120 kDa.
  • Supplied as a 5 μg aliquot.
Catalog No. 50-002-91277 Supplier Medchemexpress LLC Supplier No. HYP754045UG
Recombinant mouse CD43 (leukosialin) extracellular domain produced in HEK293 cells and supplied as an Fc fusion protein. CD43 plays a regulatory role in T cell activation, proliferation, differentiation, trafficking, and migration. The product is supplied as a 5 μg aliquot with reported purity of 95% and an apparent molecular weight of ~72-120 kDa.
  • Recombinant extracellular domain of mouse CD43.
  • Expressed in HEK293 cells for mammalian post-translational modifications.
  • Fc fusion tag for improved stability and detection.
  • Reported purity of 95.0%.
  • Apparent molecular weight approximately 72-120 kDa.
  • Supplied as a 5 μg aliquot.