Learn More
Medchemexpress LLC CD43 protein, mouse (HEK293, Fc) | 100 UG
SDP
Catalog No. 5000291348
Encompass_Preferred
Recombinant mouse CD43 extracellular fragment (D20-G248) fused to an Fc tag, expressed in HEK293 cells and supplied as a lyophilized preparation for research use in binding, signaling, and cell biology assays.
  • Recombinant extracellular fragment spanning D20-G248.
  • Fc tag for improved stability and detection.
  • Expressed in HEK293 to preserve mammalian glycosylation.
  • Lyophilized powder, formulated in PBS, pH 7.4.
  • Low endotoxin level (<1 EU/μg); store frozen and aliquot after reconstitution.
Catalog No. 50-002-91348 Supplier Medchemexpress LLC Supplier No. HYP75404100UG
Recombinant mouse CD43 extracellular fragment (D20-G248) fused to an Fc tag, expressed in HEK293 cells and supplied as a lyophilized preparation for research use in binding, signaling, and cell biology assays.
  • Recombinant extracellular fragment spanning D20-G248.
  • Fc tag for improved stability and detection.
  • Expressed in HEK293 to preserve mammalian glycosylation.
  • Lyophilized powder, formulated in PBS, pH 7.4.
  • Low endotoxin level (<1 EU/μg); store frozen and aliquot after reconstitution.