Learn More
Medchemexpress LLC CD43 protein, mouse, recombinant (HEK293, Fc) | 10 UG
SDP
Catalog No. 5000291782
Encompass_Preferred
Recombinant mouse CD43 (sialophorin) extracellular domain produced in HEK293 cells and fused to an Fc tag. Supplied in microgram-scale quantities for use in binding studies, assay development, and detection of CD43 interactions.
  • Recombinant extracellular domain of mouse CD43.
  • Produced in HEK293 cells to retain mammalian post-translational modifications.
  • Fc fusion enables easy detection and immobilization.
  • Suitable for biochemical and cell-based assays.
  • Available in microgram quantities for small-scale experiments.
  • Reported molecular weight range 72-120 kDa.
Catalog No. 50-002-91782 Supplier Medchemexpress LLC Supplier No. HYP7540410UG
Recombinant mouse CD43 (sialophorin) extracellular domain produced in HEK293 cells and fused to an Fc tag. Supplied in microgram-scale quantities for use in binding studies, assay development, and detection of CD43 interactions.
  • Recombinant extracellular domain of mouse CD43.
  • Produced in HEK293 cells to retain mammalian post-translational modifications.
  • Fc fusion enables easy detection and immobilization.
  • Suitable for biochemical and cell-based assays.
  • Available in microgram quantities for small-scale experiments.
  • Reported molecular weight range 72-120 kDa.