Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC CD43 protein, rat (HEK293, Fc) | 95.0% | 100 UG
SDP

Catalog No. 5000293423
Encompass_Preferred

Recombinant rat CD43 Fc-fusion protein expressed in HEK293 cells for research use in immunology and cell-signaling assays. The construct corresponds to residues Q21-S243 of the rat CD43 sequence and includes a C-terminal human Fc to facilitate detection and immobilization.

  • Species: rat.
  • Expression system: HEK293 cells.
  • Tag: C-terminal human Fc fusion.
  • Sequence region: Q21-S243 (accession R9PXZ7).
  • Purity: 95.0% (determined by reducing SDS-PAGE).
  • Recommended reconstitution concentration: < 100 μg/mL.
  • Intended use: research applications in immunology and cell signaling.

Catalog No. 50-002-93423 Supplier Medchemexpress LLC Supplier No. HYP75403100UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant rat CD43 Fc-fusion protein expressed in HEK293 cells for research use in immunology and cell-signaling assays. The construct corresponds to residues Q21-S243 of the rat CD43 sequence and includes a C-terminal human Fc to facilitate detection and immobilization.

  • Species: rat.
  • Expression system: HEK293 cells.
  • Tag: C-terminal human Fc fusion.
  • Sequence region: Q21-S243 (accession R9PXZ7).
  • Purity: 95.0% (determined by reducing SDS-PAGE).
  • Recommended reconstitution concentration: < 100 μg/mL.
  • Intended use: research applications in immunology and cell signaling.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.