Learn More
Medchemexpress LLC CD43 protein, rat (HEK293, Fc) | >95.0% | 5 UG
SDP
Catalog No. 5000291720
Encompass_Preferred
Recombinant rat CD43 extracellular domain fused to an Fc tag, expressed in HEK293 cells and supplied as a lyophilized protein for biochemical and immunological assays. Certificate of analysis reports >95% purity by reducing SDS-PAGE and a glycosylation-dependent molecular weight range. Formulated in PBS and recommended to be reconstituted at ≥100 μg/mL; store frozen for long-term stability.
Catalog No. 50-002-91720 Supplier Medchemexpress LLC Supplier No. HYP754035UG
Recombinant rat CD43 extracellular domain fused to an Fc tag, expressed in HEK293 cells and supplied as a lyophilized protein for biochemical and immunological assays. Certificate of analysis reports >95% purity by reducing SDS-PAGE and a glycosylation-dependent molecular weight range. Formulated in PBS and recommended to be reconstituted at ≥100 μg/mL; store frozen for long-term stability.