Learn More
Biocare Medical, LLC CD44 (HCAM) Antibody - Concentrate, 0.1 ml
SDP
Supplier:  Biocare Medical, LLC CM318A
Encompass_Preferred
CD44 is a transmembranous glycoprotein (80 kDa) responsible for cell adhesion that is expressed in a wide range of normal tissues, such as tonsil, breast, prostate, skin, bladder and cervical squamous epithelium (1). Studies have shown that CD44, also known as HCAM, plays an important role in the cascade of metastasis and progression of human malignant tumors (2). In breast cancer studies, evaluation of CD44 expression has been useful in the differentiation of benign and malignant papillary lesions (2). Expression of CD44s, the most common CD44 isoform, has been reported to be associated with increased survival of patients with nodenegative invasive breast carcinoma (3). Further studies have shown a subpopulation of CD44 /CD24- in breast cancer cells have stem/progenitor cell properties (4).
Catalog No. 50-826-41