Learn More
enQuire Bioreagents Calponin-1 Anti-Human Monoclonal [Clone SPM169], Size: 100ug
Supplier:  enQuire Bioreagents 960MSM1P1
Encompass_Preferred
Product Type: Antibody. Application: Flow, IF, WB, IHC. Conjugate/Tag: Unconjugated. Clone: 156-3C11. Clonality: Monoclonal. Host: Mouse. Isotype: IgG2a kappa. Positive Control: HeLa cells or paracortex in tonsil or lymph node. Cellular Staining: Cell Surface. Buffer: 10mM PBS with 0.05% BSA & 0.05% azide. Concentration: 200ug/ml. Purity: Protein A/G Purified. Immunogen: Stimulated human leukocytes. Storage: Short term: Store at 2-4C. Long term, aliquot and store at -20C. MW: 80-95kDa.Chromosome: 11p13. Gene Symbol: CD44. Gene ID: 960. Ensemble: ENSG00000026508. Uniprot: XP_016874073.
Catalog No. 50-188-5355