Learn More
enQuire Bioreagents Calponin-1 Anti-Human Monoclonal [Clone SPM169], Size: 100ug
Supplier:  enQuire Bioreagents 960MSM2P1
Encompass_Preferred
Product Type: Antibody. Application: Flow, IF, IHC. Additionally, this antibody has been validated by MS-QAVA(TM). Specificity has been quantitatively determined via Mass Spec. Conjugate/Tag: Unconjugated. Clone: HCAM/918. Clonality: Monoclonal. Host: Mouse. Isotype: IgG2a kappa. Positive Control: DU145 cells. Tonsil, lymph node or breast carcinomas. Cellular Staining: Cell surface. Buffer: 10mM PBS with 0.05% BSA & 0.05% azide. Concentration: 200ug/ml. Purity: Protein A/G Purified. Immunogen: Recombinant human HCAM protein. Storage: Short term: Store at 2-4C. Long term, aliquot and store at -20C. MW: 80-95kDa.Chromosome: 11p13. Gene Symbol: CD44. Gene ID: 960. Ensemble: ENSG00000026508. Uniprot: XP_016874073.
Catalog No. 50-188-5358