Learn More
Aviva Systems Biology Corp CD44 Antibody - C-terminal region (ARP61023_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP61023_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CD44. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the C terminal region of human CD44. Alternate names: CDW44, CSPG8, ECMR-III, HCELL, IN, LHR, MC56, MDU2, MDU3, MGC10468, MIC4, MUTCH-I, Pgp1, HUTCH-IApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-287-234